1-Heptene, 98+%, Thermo Scientific Chemicals
1-Heptene, 98+%, Thermo Scientific Chemicals
1-Heptene, 98+%, Thermo Scientific Chemicals
1-Heptene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Heptene, 98+%, Thermo Scientific Chemicals
CAS: 592-76-7 | C7H14 | 98.189 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11487.AE100 mL
Catalog number ALFL11487.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS575-03-1
IUPAC Name7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one
Molecular FormulaC10H5F3O3
InChI KeyCCKWMCUOHJAVOL-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)Pale cream or pale pink to pink
Melting Point (clear melt)182.0-188.0?C