trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-1,2-Cyclohexanedicarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 2305-32-0 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL11288.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10112-91-1
IUPAC Namedimercuranebis(ylium) dichloride
Molecular FormulaCl2Hg2
InChI KeyZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg+][Hg+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Titration Iodimetric>=99.0 %
Mercuric Chloride=<0.01 % (HgCl2)
Appearance (Form)Powder
Residue after reduction=<0.02 %