2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
CAS: 2285-12-3 | C8H4F3NO | 187.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11156.1425 g
Catalog number ALFL11156.14
Price (MYR)
1,193.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,193.00
Add to cart
Specifications
Chemical Name or Material2-(Trifluoromethyl)phenyl isocyanate
CAS2285-12-3
Health Hazard 1H226-H312-H315-H319-H330-H334-H335
Health Hazard 2GHS H Statement
H330-H334-H226-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
View more
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only
General References:
  1. Kevin G Liu.; Millard H Lambert.; Lisa M Leesnitzer.; William Oliver Jr.; Ronda J Ott.; Kelli D Plunket.; Ludwig W Stuart.; Peter J Brown.; Timothy M Willson.; Daniel D Sternbach. Identification of a series of PPARγ/δ dual agonists via solid-Phase parallel synthesis. Bioorganic & Medicinal Chemistry Letters. 2001, 11 (22), 2959-2962.
  2. Zhaoguang Li.; He Huang.; Hongbin Sun.; Hualiang Jiang.; Hong Liu. Microwave-Assisted Efficient and Convenient Synthesis of 2,4(1H,3H)-Quinazolinediones and 2-Thioxoquinazolines. J. Comb. Chem. 2008, 10 (3), 484-486.