2,4-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichlorophenyl isocyanate, 96%, Thermo Scientific Chemicals
CAS: 2612-57-9 | C7H3Cl2NO | 188.007 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11089.065 g
Catalog number ALFL11089.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dichlorophenyl isocyanate
CAS2612-57-9
Health Hazard 1H301+H311+H331-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c