4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
CAS: 2131-61-5 | C7H4N2O2S | 180.181 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11042.1850 g
Catalog number ALFL11042.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS31410-01-2
IUPAC Name1-(prop-2-en-1-yl)-1H-imidazole
Molecular FormulaC6H8N2
InChI KeyXLXCHZCQTCBUOX-UHFFFAOYSA-N
SMILESC=CCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to white to yellow
Refractive Index1.5025-1.5060 @ 20?C
FormLiquid
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
Solubility
It hydrolyzes with water. Soluble in toluene.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
  1. M Péter.; A Gyeresi.; F Fülöp. Liquid chromatographic enantioseparation of β-blocking agents with (1R, 2R)-1, 3-diacetoxy-1-(4-nitrophenyl)-2-propyl isothiocyanate as chiral derivatizing agent. Journal of Chromatography A. 2001, 910,(2), 247-253.
  2. YJ Kim.; H Kwak.; SJ Lee.; JS Lee.; HJ Kwon.; SH Nam. Urea/thiourea-based colorimetric chemosensors for the biologically important ions: efficient and simple sensors. Tetrahedron. 2006, 62,(41), 9635-9640.