2-Amino-5-methylthiazole, 98+%
2-Amino-5-methylthiazole, 98+%
2-Amino-5-methylthiazole, 98+%
2-Amino-5-methylthiazole, 98+%
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98+%
CAS: 7305-71-7 | C4H6N2S | 114.166 g/mol
Have Questions?
Catalog NumberQuantity
ALFL11039.065 g
Catalog number ALFL11039.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength7.984 to 8.016 N (20°C)
Appearance (Form)Solution
Composition448 g/l (g KOH/l H20)