4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl palmitate, 98+%, Thermo Scientific Chemicals
CAS: 1492-30-4 | C22H35NO4 | 377.525 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL10896.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH59.0 to 63.0 %
Iron (Fe)=<5 ppm
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Chloride (Cl)=<10 ppm
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.
Solubility
Insoluble in water. Soluble in CHCl3.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C
RUO – Research Use Only
General References:
  1. E. Ranjbakhsh,; A.K. Bordbar,; M. Abbasia,; A.R. Khosropoura,; E. Shams. Supuran. Enhancement of stability and catalytic activity of immobilized lipase on silica-coated modified magnetite nanoparticles. Chemical Engineering Journal. 2012, 179 272-276.
  2. C K Tu, H G Thomas,; G C Wynns,; D N Silverman. Hydrolysis of 4-nitrophenyl acetate catalyzed by carbonic anhydrase III from bovine skeletal muscle. The Journal of Biological Chemistry, . 1986 , 261 10100-10103..