1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals
1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals
1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals
1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Pyrenecarboxaldehyde, 98+%, Thermo Scientific Chemicals
CAS: 3029-19-4 | C17H10O | 230.266 g/mol
Have Questions?
Catalog NumberQuantity
ALFL10835.065 g
Catalog number ALFL10835.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14191-95-8
IUPAC Name2-(4-hydroxyphenyl)acetonitrile
Molecular FormulaC8H7NO
InChI KeyAYKYOOPFBCOXSL-UHFFFAOYSA-N
SMILESOC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to dark cream or yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
Melting Point (clear melt)64.0-74.5°C
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. P. Lianos.; G. Cremel. ENVIRONMENTAL EFFECTS ON THE ELECTRONIC SPECTRAL PROPERTIES OF 1-PYRENECARBOXALDEHYDE AND THEIR APPLICATION IN PROBING BIOLOGICAL STRUCTURES. Photochemistry and Photobiology. 1980, 31 (5), 429-434.
  2. Fernando Secco.; Marcella Venturin.; Tarita Biver.;Francisco Sánchez.; Rafael Prado-Gotor.; Elia Grueso. Solvent Effects on the Kinetics of the Interaction of 1-Pyrenecarboxaldehyde with Calf Thymus DNA. J. Phys. Chem. B. 2010, 114 (13), 4686-4691.