(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFL10294.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS141-79-7
IUPAC Name4-methylpent-3-en-2-one
Molecular FormulaC6H10O
InChI KeySHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILESCC(C)=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC90 % alpha-Isomer
Refractive index1.4420 to 1.4460 (20°C, 589 nm)
Density(20/4°C) 0.852 to 0.856 kg/l
View more
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.
Solubility
Soluble in water. (100 g/L)
Notes
Light sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
RUO – Research Use Only
General References:
  1. K-P. Wilhelm.; B. Böttjer.; C-P. Siegers. Quantitative assessment of primary skin irritants in vitro in a cytotoxicity model: comparison with in vivo human irritation tests.British Journal of Dermatology. 2001, 145 (5),709-715 .
  2. D. Huang.; Y.J. Wang.; Y.C. Cui.; G.S. Luo. Direct synthesis of mesoporous TiO2 and its catalytic performance in DBT oxidative desulfurization.Microporous Mesoporous Mater. 2008, 116 (1-3),378-385 .