(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1-Tetradecyl)trimethylammonium bromide, 98%, Thermo Scientific Chemicals
CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFL10294.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS51-17-2
IUPAC Name1H-1,3-benzodiazole
Molecular FormulaC7H6N2
InChI KeyHYZJCKYKOHLVJF-UHFFFAOYSA-N
SMILESN1C=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point171°C to 174°C
HPLC>=97.5 %
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder or crystals
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.
Solubility
Soluble in water. (100 g/L)
Notes
Light sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
RUO – Research Use Only
General References:
  1. K-P. Wilhelm.; B. Böttjer.; C-P. Siegers. Quantitative assessment of primary skin irritants in vitro in a cytotoxicity model: comparison with in vivo human irritation tests.British Journal of Dermatology. 2001, 145 (5),709-715 .
  2. D. Huang.; Y.J. Wang.; Y.C. Cui.; G.S. Luo. Direct synthesis of mesoporous TiO2 and its catalytic performance in DBT oxidative desulfurization.Microporous Mesoporous Mater. 2008, 116 (1-3),378-385 .