L-Lactide, 98+%, Thermo Scientific Chemicals
L-Lactide, 98+%, Thermo Scientific Chemicals
L-Lactide, 98+%, Thermo Scientific Chemicals
L-Lactide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-Lactide, 98+%, Thermo Scientific Chemicals
CAS: 4511-42-6 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL09031.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Lactide
pH3.0
Melting Point95°C to 99°C
CAS4511-42-6
Health Hazard 1H315-H319-H335
View more
L-Lactide upon polymerization gives poly(lactic acid), which finds many useful properties like biodegradable and nontoxic to the human body and the environment. Its polymer, poly-l-lactide is utilized in the repairing of orbital posterior floor fracture. Further, it is used as biomedical materials for tissue regeneration, matrices for drug delivery systems and alternatives for commercial polymeric materials to reduce the impact on the environment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in chloroform, and methanol. Slightly soluble in benzene.
Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Coulembier, O.; Moins, S.; Maji, S.; Zhang, Z.; De Geest, B. G.; Dubois, P.; Hoogenboom, R. Linear polyethylenimine as (multi) functional initiator for organocatalytic l-lactide polymerization. J. Mater. Chem. B 2015, 3 (4), 612-619.
  2. Kober, E.; Petrus, R.; Kocięcka, P.; Janas, Z.; Sobota, P. Lithium diaminebis(aryloxido) complexes: Synthesis, structures and reactivity in l-lactide polymerization. Polyhedron 2015, 85, 814-823.