Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
CAS: 127-63-9 | C12H10O2S | 218.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFL08232.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiphenyl sulfone
CAS127-63-9
Health Hazard 1H302
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Murafuji T, et al. Heterocyclic bismuth carboxylates based on a diphenyl sulfone scaffold: synthesis and antifungal activity against Saccharomyces cerevisiae.European J. Med. Chem.,2013,63531-535.
  2. A. Apicella, et al. Thermokinetics and chemorheology of the cure reactions of the tetraglycidyl diamino diphenyl methane-diamino diphenyl sulfone epoxy systems.J. Appl. Polym. Sci.,1984,29(6), 2083-2096.