4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL08200.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS28611-39-4
IUPAC Name[4-(dimethylamino)phenyl]boronic acid
Molecular FormulaC8H12BNO2
InChI KeyRIIPFHVHLXPMHQ-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Appearance (Form)Powder and/or lumps
Infrared spectrumConforms
Titration with NaOH>=95 %
4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.
Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Mukherjee, S.; Thilagar, P. Recent advances in purely organic phosphorescent materials. Chem. Commun. 2015, 51 (55), 10988-11003.
  2. Ravoet, J.; Reybroeck, W.; De Graaf, D. C. Pesticides for Apicultural and/or Agricultural Application Found in Belgian Honey Bee Wax Combs. B. Environ. Contam. Tox. 2015, 94 (5), 543-548.