2-Methyl-3-pentanol, 97+%, Thermo Scientific Chemicals
2-Methyl-3-pentanol, 97+%, Thermo Scientific Chemicals
2-Methyl-3-pentanol, 97+%, Thermo Scientific Chemicals
2-Methyl-3-pentanol, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyl-3-pentanol, 97+%, Thermo Scientific Chemicals
CAS: 565-67-3 | C6H14O | 102.177 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08015.0910 g
Catalog number ALFL08015.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-3-pentanol
CAS565-67-3
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-3-pentanol was used as a volatile marker. It used as a fuel.
Solubility
Amalgamate with ethanol, ethyl ether, benzene. Insoluble in water
Notes
Incompatible materials are oxidizing agents, acids, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only
Angelo C Salvador; Inês Baptista; António S Barros; Newton C M Gomes; Angela Cunha; Adelaide Almeida; Silvia M Rocha. Can volatile organic metabolites be used to simultaneously assess microbial and mite contamination level in cereal grains and coffee beans?PLoS ONE.1968, 8 (4), e59338.