Acetazolamide, 99%, Thermo Scientific Chemicals
Acetazolamide, 99%, Thermo Scientific Chemicals
Acetazolamide, 99%, Thermo Scientific Chemicals
Acetazolamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetazolamide, 99%, Thermo Scientific Chemicals
CAS: 59-66-5 | C4H6N4O3S2 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL07562.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetazolamide
CAS59-66-5
Health Hazard 1H360
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
RUO – Research Use Only
General References:
  1. Junwei Gao; Xiaohua Wang; Yongjie Chang; Jianzhao Zhang; Qianliu Song; Heming Yu; Xuejun Li. Acetazolamide inhibits osmotic water permeability by interaction with aquaporin-1.Analytical Biochemistry.2006, 350 (2), 165-170 .
  2. S. Nigel Cox, FRCS; Elizabeth Hay, MD; Alan C. Bird, MD. Treatment of Chronic Macular Edema With Acetazolamide.Arch Ophthalmol.1988, 106 (9), 1190-1195.