Methyl linoleate, 95%, Thermo Scientific Chemicals
Methyl linoleate, 95%, Thermo Scientific Chemicals
Methyl linoleate, 95%, Thermo Scientific Chemicals
Methyl linoleate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl linoleate, 95%, Thermo Scientific Chemicals
CAS: 112-63-0 | C19H34O2 | 294.479 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL07450.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1066-50-8
IUPAC Nameethylphosphonoyl dichloride
Molecular FormulaC2H5Cl2OP
InChI KeyOWGJXSYVHQEVHS-UHFFFAOYSA-N
SMILESCCP(Cl)(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4645-1.4685 @ 20?C
FormLiquid
Assay (GC)≥97.5%
The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.
Solubility
Soluble in DMF, fat solvents, oils.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Marcus. Karel.; Karen. Schaich.; Ram B. Roy. Interaction of peroxidizing methyl linoleate with some proteins and amino acids. J. Agric. Food Chem. 1975, 23, (2), 159-163.
  2. Yasunobu Shimada.; Yrjo Roos.; Marcus Karel. Oxidation of methyl linoleate encapsulated in amorphous lactose-based food model. J. Agric. Food Chem. 1991, 39, (4), 637-641.