Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 531-81-7 | C10H6O4 | 190.154 g/mol
Have Questions?
Catalog NumberQuantity
ALFL07133.1425 g
Catalog number ALFL07133.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4480-83-5
IUPAC Name1,4-dioxane-2,6-dione
Molecular FormulaC4H4O4
InChI KeyPIYNUZCGMLCXKJ-UHFFFAOYSA-N
SMILESO=C1COCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting point90°C to 95°C
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
Titration excess NaOH>=96.0 %
Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water (13 g/L at 37°C) and alcohol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Yefim Manevich; Kathryn D.Held; John E.Biaglow. Coumarin-3-carboxylic acid as a detector for hydroxyl radicals generated chemically and by gamma radiation. Radiation Research. 1997, 148,(6), 580-591.
  2. Tz. Mihaylov; N.Trendafilova; I.Kostova; I.Georgieva; G.Bauer. DFT modeling and spectroscopic study of metal-ligand bonding in La(III) complex of coumarin-3-carboxylic acid. Chemical Physics. 2006, 327,(2-3), 209-219.