3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
3-Benzyloxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFL06830.0910 g
Catalog number ALFL06830.09
Price (MYR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Benzyloxy-4-methoxybenzaldehyde
CAS6346-05-0
Melting Point60°C to 64°C
Recommended StorageAmbient temperatures
EINECS Number228-752-7