1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%, Thermo Scientific Chemicals
CAS: 303180-11-2 | C5H6N2O5 | 174.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL06731.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS15231-91-1
IUPAC Name6-bromonaphthalen-2-ol
Molecular FormulaC10H7BrO
InChI KeyYLDFTMJPQJXGSS-UHFFFAOYSA-N
SMILESOC1=CC=C2C=C(Br)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Infrared spectrumConforms
Melting point125°C to 130°C
GC>=96.0 %
Appearance (Form)Powder