Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium 1-nonanesulfonate, 98+%, Thermo Scientific Chemicals
CAS: 35192-74-6 | C9H19NaO3S | 230.298 g/mol
Have Questions?
Catalog NumberQuantity
ALFL06407.065 g
Catalog number ALFL06407.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium 1-nonanesulfonate
CAS35192-74-6
Melting Point>300°C
Recommended StorageAmbient temperatures
TSCANo
View more
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Tyagi, M. G.; Mathew, S. K. Lipoprotein associated phospholipase A2 enzyme; possible new roles and inhibition for therapeutic intervention. Int. J. Res. Med. Sci. 2014, 2 (3), 805-809.
  2. Pöllmann, H.; Stefan, S.; Stern, E. Synthesis, characterization and reaction behaviour of lamellar AFm phases with aliphatic sulfonate-anions. Cem. Concr. Res. 2006, 36 (11), 2039-2048.