3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL05948.22
Price (MYR)
679.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
679.00
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzotrifluoride
CAS98-46-4
Health Hazard 1H227-H302+H312-H315-H319-H330-H335
Health Hazard 2GHS H Statement
H301-H311-H330-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c