3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrobenzotrifluoride, 97%, Thermo Scientific Chemicals
CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFL05948.14
Price (MYR)
338.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
338.00
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzotrifluoride
CAS98-46-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H311-H330-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.