Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
CAS: 100-41-4 | C8H10 | 106.168 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number ALFL05908.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS174899-66-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear, colorless or yellow to red/brown
Assay (unspecified)>97.5%
Refractive Index1.4370-1.4410 @ 20?C
Water Content (Karl Fischer Titration)<0.5%
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.
Solubility
Miscible with alcohol, ethylalcohol, benzene, ethyl ether, carbon tetrachloride, organic solvents, sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Smith, R. S.; May, R. A.; Kay, B. D. Probing Toluene and Ethylbenzene Stable Glass Formation Using Inert Gas Permeation. J. Phys. Chem. Lett. 2015, 6 (18), 3639-3644.
  2. Yang, L.; Fan, B.; Cui, X.; Shi, X.; Li, R. Solvent-free aerobic oxidation of ethylbenzene over Mn-containing silylated MgAl layered double hydroxides. J. Ind. Eng. Chem. 2015, 21, 689-695.