Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylbenzene, 99%, Thermo Scientific Chemicals
CAS: 100-41-4 | C8H10 | 106.168 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
100 mL
500 mL
Catalog number ALFL05908.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Chemical Identifiers
CAS1308-06-1
IUPAC Namedicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular FormulaCo3O4
InChI KeyUBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1% (U.S. specification)
Appearance (Color)Fine grey to black
Complexometric TitrationCo content: 71-74% (Theory 73.42%, non-U.S. specification)
FormPowder
CommentMaterial specification is different for U.S. and non-U.S. purchases
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.
Solubility
Miscible with alcohol, ethylalcohol, benzene, ethyl ether, carbon tetrachloride, organic solvents, sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Smith, R. S.; May, R. A.; Kay, B. D. Probing Toluene and Ethylbenzene Stable Glass Formation Using Inert Gas Permeation. J. Phys. Chem. Lett. 2015, 6 (18), 3639-3644.
  2. Yang, L.; Fan, B.; Cui, X.; Shi, X.; Li, R. Solvent-free aerobic oxidation of ethylbenzene over Mn-containing silylated MgAl layered double hydroxides. J. Ind. Eng. Chem. 2015, 21, 689-695.