Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads, Thermo Scientific Chemicals
Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads, Thermo Scientific Chemicals
Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads, Thermo Scientific Chemicals
Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads, Thermo Scientific Chemicals
CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number ALFL05335.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Solubility InformationInsoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
Unit Size
Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.