2-(Ethylthio)ethanol, 96%, Thermo Scientific™
2-(Ethylthio)ethanol, 96%, Thermo Scientific™
2-(Ethylthio)ethanol, 96%, Thermo Scientific™
2-(Ethylthio)ethanol, 96%, Thermo Scientific™
2-(Ethylthio)ethanol, 96%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFL05040.1850 g
Catalog number ALFL05040.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2-(Ethylthio)ethanol
CAS110-77-0
Health Hazard 1H227-H318
Health Hazard 2GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P280-P305+P351+P338-P310-P370+P378q-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.