2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals
2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals
2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals
2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals
CAS: 622-24-2 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFL04891.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1128-08-1
IUPAC Name3-methyl-2-pentylcyclopent-2-en-1-one
Molecular FormulaC11H18O
InChI KeyYCIXWYOBMVNGTB-UHFFFAOYSA-N
SMILESCCCCCC1=C(C)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4780-1.4820 @ 20?C
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow