(R)-(+)-Limonene, 96%, Thermo Scientific Chemicals
(R)-(+)-Limonene, 96%, Thermo Scientific Chemicals
(R)-(+)-Limonene, 96%, Thermo Scientific Chemicals
(R)-(+)-Limonene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(R)-(+)-Limonene, 96%, Thermo Scientific Chemicals
CAS: 5989-27-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL04733.AP
Price (MYR)
414.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
414.00
Add to cart
Specifications
Chemical Name or Material(R)-(+)-Limonene
Name Notestabilized
CAS5989-27-5
Health Hazard 1H226-H304-H315-H317-H319
Health Hazard 2GHS H Statement
H226-H315-H317
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
View more
Chiral building block. Biodegradable solvent. Limonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Limonene is used as a solvent for filament-fused 3D printing, in cosmetic products, as a fragrance and as a biofuel. It is useful to promote weight loss, prevent cancer, treat cancer and treat bronchitis. It also finds use in foods, beverages, chewing gum, as a flavoring agent and a shrinking agent to dissolve polystyrene. It acts as a chiral intermediate for natural product synthesis and as an insect repellant.
Solubility
Miscible with alcohol,chloroform,ether and carbondisulfide.Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only
General References:
  1. Chiral building block. Biodegradable solvent.
  2. Bedjanian, Y.; Romanias, M. N.; Morin, J. Reaction of Limonene with F2: Rate Coefficient and Products. J. Phys. Chem. A. 2014, 118 (44), 10233-10239.
  3. Faure, K.; Bouju, E.; Suchet, P.; Berthod, A. Use of Limonene in Countercurrent Chromatography: A Green Alkane Substitute. Anal. Chem. 2013, 85 (9), 4644-4650.
  4. Auriemma, F.; Rosa, C. D.; Caprio,M. R. D.; Girolamo, R. D.; Coates, G. W. Crystallization of Alternating Limonene Oxide/Carbon Dioxide Copolymers: Determination of the Crystal Structure of Stereocomplex Poly(limonene carbonate). Macromolecules 2015, 48 (8), 2534-2550.