N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFL04694.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS120-71-8
IUPAC Name2-methoxy-5-methylaniline
Molecular FormulaC8H11NO
InChI KeyWXWCDTXEKCVRRO-UHFFFAOYSA-N
SMILESCOC1=CC=C(C)C=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to black
Appearance (Form)Crystalline chunks
Infrared spectrumConforms
Melting point49°C to 55°C
GC>=94 %
N,N'-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.
Solubility
Soluble in water, ethanol and methanol.
Notes
Air and light sensitive. Store in a cool place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron, iron salts, copper, aluminum, brass and free radical initiators.
RUO – Research Use Only
General References:
  1. Graiff, C.; Pontiroli, D.; Bergamonti, L.; Cavallari, C.; Lottici, P. P.; Predieri, G. Structural investigation of N,N'-methylenebisacryl­amide via X-ray diffraction assisted by crystal structure prediction. J. Appl. Crystallogr. 2015, 48, 550-557.
  2. Lazzari, S.; Pfister, D.; Diederich, V.; Kern, A.; Storti, G. Modeling of Acrylamide,N'-Methylenebisacrylamide Solution Copolymerization. Ind. Eng. Chem. Res. 2014, 53 (22), 9035-9048.