Triethanolamine, 98+%, Thermo Scientific Chemicals
Triethanolamine, 98+%, Thermo Scientific Chemicals
Triethanolamine, 98+%, Thermo Scientific Chemicals
Triethanolamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triethanolamine, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFL04486.22100 g
Catalog number ALFL04486.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=99.0 %
pH7.9 to 8.3 (5 % at 20°C)
Nitrate (NO3)=<100 ppm
Potassium (K)=<100 ppm
View more
Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating. Further, it is used in hair care products like hair dyes and wave sets.
Solubility
Miscible with water, methanol, acetone, ether, carbon tetrachloride, n-heptane and chloroform. Slightly soluble in petroleum ether.
Notes
Air sensitive and hygroscopic Incompatible with acids, oxidizing agents, reducing agents and metals.
RUO – Research Use Only
General References:
  1. Reacts with N-alkylanilines in dioxane (autoclave) in the presence of Dichlorotris(triphenyl phosphine) ruthenium(II) , L00373, to give 1-alkyl indoles in good yield: Synth. Commun., 26, 1349 (1996).
  2. Du, M.; Yin, X.; Gong, H. Effects of triethanolamine on the morphology and phase of chemically deposited tin sulfide. Mater. Lett. 2015, 152, 40-44.
  3. Chowdhury, P.; Gomaa, H.; Ray, A. K. Sacrificial hydrogen generation from aqueous triethanolamine with Eosin Y-sensitized Pt/TiO2 photocatalyst in UV, visible and solar light irradiation. Chemosphere 2015, 121, 54-61.