4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
CAS: 637-89-8 | C6H6OS | 126.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04429.065 g
Catalog number ALFL04429.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS16387-71-6
IUPAC Namedeca-4,6-diyne
Molecular FormulaC10H14
InChI KeyLIWZSNTUMSGWTF-UHFFFAOYSA-N
SMILESCCCC#CC#CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to brown
FormLiquid
Assay from Supplier's CofA≥96.0%
4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures
Solubility
Soluble in water.
Notes
Air Sensitive. Store at 4°C. Incompatible with air and oxidizing agents.
RUO – Research Use Only
General References:
  1. Shaowei Chen. 4-Hydroxythiophenol-Protected Gold Nanoclusters in Aqueous Media.J. Am. Chem. Soc.1999, 15 (22), 7551-7557.
  2. Hiroshi Sugiyama; Tsuyoshi Fujiwara; Isao Saito. Activation of neocarzinostatin-chromophore by 4-hydroxythiophenol: Intramolecular radical trapping of biradical intermediate.Tetrahedron Letters.1994, 35 8825-8828.