1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals
1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals
1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals
1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,12-Diaminododecane, 98+%, Thermo Scientific Chemicals
CAS: 2783-17-7 | C12H28N2 | 200.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL04258.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,12-Diaminododecane
CAS2783-17-7
Health Hazard 1H302-H314-H317-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.
Solubility
Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only
General References:
  1. A Sintov; N Di-Capua; A Rubinstein. Cross-linked chondroitin sulphate: characterization for drug delivery purposes.Biomaterials.1995, 16 (6), 473-478.
  2. Anthony T Iavarone; Evan R Williams. Mechanism of charging and supercharging molecules in electrospray ionization.Journal of the American Chemical Society.2003, 125 (8), 2319-2327.