View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfamerazine
CAS127-79-7
Melting Point234°C to 238°C
Recommended StorageAmbient temperatures
RTECS NumberWP0750000