2-Ethylhexyl acrylate, 98%, stab., Thermo Scientific Chemicals
2-Ethylhexyl acrylate, 98%, stab., Thermo Scientific Chemicals
2-Ethylhexyl acrylate, 98%, stab., Thermo Scientific Chemicals
2-Ethylhexyl acrylate, 98%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Ethylhexyl acrylate, 98%, stab., Thermo Scientific Chemicals
CAS: 103-11-7 | C11H20O2 | 184.279 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number ALFL03854.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS917-64-6
IUPAC Nameiodo(methyl)magnesium
Molecular FormulaCH3IMg
InChI KeyAUPXBVDHVRZMIB-UHFFFAOYSA-M
SMILESC[Mg]I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear to hazy solution
Strength2.85 to 3.50 M
Appearance (Color)Yellow to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.
Solubility
Miscible with ether and alcohol. Immiscible with water.
Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Zhang, Y.; Dubé, M. A. Copolymerization of 2-Ethylhexyl Acrylate and D-Limonene. Polym. Plast. Technol. Eng. 2015, 54 (5), 499-505.
  2. Kockler, K. B.; Haehnel, A. P.; Fleischhaker, F.; Schneider-Baumann, M.; Misske, A. M.; Barner-Kowollik, C. No Apparent Correlation of kp with Steric Hindrance for Branched Acrylates. Macromol. Chem. Phys. 2015, 216 (14), 1573-1582.