N,N-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
N,N-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
N,N-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
N,N-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals
CAS: 109-55-7 | C5H14N2 | 102.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
Catalog number ALFL03539.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Dimethyl-1,3-propanediamine
CAS109-55-7
Health Hazard 1H226-H302+H312-H314-H317-H335
Health Hazard 2GHS H Statement
H314-H226-H302-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
View more
Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to methyl hydrazine. DMAPA is also used directly as a hardener in epoxy resins in the plastics industry, as a cross linking agent for cellulose fibers in the paper industry and as an anti-shriking agent for leather. It is used as an intermediate in the production of binding agents, ion-exchange materials, flocculating agents (water treatment), cosmetic agents, washing and cleaning agents (betaines), additive for petrol and other fuels, polyurethane fibers and lubricants, dyes, agrochemicals, agents used in the photographic and textile industries.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
  1. Sofie Meng Andersen; Mads Heuckendorff; Henrik H Jensen. 3-(Dimethylamino)-1-propylamine: a cheap and versatile reagent for removal of byproducts in carbohydrate chemistry. Organic Letters. 2015, 17, (4), 944-947.
  2. Marcos Hervella; Juan I Yanguas; María E Z Iglesias; Mónica Larrea; Concepción Ros; Manuel Gallego. [Contact allergy to 3-dimethylaminopropylamine and cocamidopropyl betaine]. Actas Dermosifiliogr. 2006, 97, (3), 189-195.
  3. Useful for cleavage of N-alkylphthalimide derivatives to give primary amines, as an alternative to the carcinogenic methylhydrazine: Org. Synth. Coll., 9, 13, 16 (1993).