4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
CAS: 5434-21-9 | C8H7NO4 | 181.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL03333.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS93-61-8
IUPAC NameN-methyl-N-phenylformamide
Molecular FormulaC8H9NO
InChI KeyJIKUXBYRTXDNIY-UHFFFAOYSA-N
SMILESCN(C=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Refractive index1.5600 to 1.5620 (20°C, 589 nm)