4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophthalic acid, 98%, Thermo Scientific Chemicals
CAS: 5434-21-9 | C8H7NO4 | 181.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL03333.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA>99.96% (metals basis)
Assay from Supplier's CofAAg < 0.008%
Assay from Supplier's CofACu < 0.005%
Assay from Supplier's CofAFe < 0.005%