4-Methoxychalcone, 97%, Thermo Scientific Chemicals
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxychalcone, 97%, Thermo Scientific Chemicals
CAS: 959-33-1 | C16H14O2 | 238.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL03319.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methoxychalcone
CAS959-33-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point73°C to 76°C