4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
CAS: 6373-50-8 | C16H12Cl2N4OS | 379.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL03184.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Cyclohexylaniline
CAS6373-50-8
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Mou, J.; Park, A.; Cai, Y.; Yuan, J.; Yuan, C. Structure-activity relationship study of E6 as a novel necroptosis inducer. Bioorg. Med. Chem. Lett. 2015, 25 (15), 3057-3061.
  2. Wei, W.; Wen, J.; Yang, D.; Sun, X.; You, J.; Suo, Y.; Wang, H. Iron-catalyzed three-component tandem process: a novel and convenient synthetic route to quinoline-2,4-dicarboxylates from arylamines, glyoxylic esters, and alpha-ketoesters. Tetrahedron 2013, 69 (50), 10747-10751.