5-Aminonaphthalene-2-sulfonic acid, 97%, Thermo Scientific Chemicals
5-Aminonaphthalene-2-sulfonic acid, 97%, Thermo Scientific Chemicals
5-Aminonaphthalene-2-sulfonic acid, 97%, Thermo Scientific Chemicals
5-Aminonaphthalene-2-sulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aminonaphthalene-2-sulfonic acid, 97%, Thermo Scientific Chemicals
CAS: 119-79-9 | C10H9NO3S | 223.246 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03099.1425 g
Catalog number ALFL03099.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS515-42-4
IUPAC Namesodium benzenesulfonate
Molecular FormulaC6H5NaO3S
InChI KeyMZSDGDXXBZSFTG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<2 % (1 g, 105°C, 3 h)