2-Chloro-2',6'-dimethylacetanilide, 99%, Thermo Scientific Chemicals
2-Chloro-2',6'-dimethylacetanilide, 99%, Thermo Scientific Chemicals
2-Chloro-2',6'-dimethylacetanilide, 99%, Thermo Scientific Chemicals
2-Chloro-2',6'-dimethylacetanilide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloro-2',6'-dimethylacetanilide, 99%, Thermo Scientific Chemicals
CAS: 1131-01-7 | C10H12ClNO | 197.662 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFL02727.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS13748-90-8
IUPAC Name(2S)-2-hydroxy-4-methylpentanoic acid
Molecular FormulaC6H12O3
InChI KeyLVRFTAZAXQPQHI-YFKPBYRVSA-N
SMILESCC(C)C[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
NMRconsistent with structure
Specific optical rotation-24° to -28° (20°C, 589 nm) (C=1, 1N NaOH)
Appearance (Color)White to off-white
Melting point77°C to 81°C
View more
2-Chloro-2',6'-dimethylacetanilide is used in the determination of the enantiomers of tocainide in blood plasma by gas-liquid chromatography with electron-capture detection. It is a nonsteroidal antinflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in Chloroform, Methanol. Insoluble in water.
Notes
Keep the container tightly closed in a cool, dry and well ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. A J Sedman; J Gal. Simultaneous determination of the enantiomers of tocainide in blood plasma using gas-liquid chromatography with electron-capture detection. Journal of Chromatography B: Biomedical Sciences and Applications. 1984, 306, 155-164.
  2. Galvez-Llompart; Maria M. Giner; Rosa C. Recio; Maria Candeletti; Sanzio Garcia-Domenech; Ramon. Application of Molecular Topology to the Search of Novel NSAIDs: Experimental Validation of Activity. Letters in Drug Design & Discovery.2010, 7, (8), 438-445.