4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.
Solubility
Miscible with water and chloroform.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only
General References:
- AlQaradawi, S. Y.; Mostafa, A.; Bazzi, H. S. Charge-transfer complexes of 4-methylpiperidine with σ- and π-acceptors. Spectrochim. Acta, Part A 2015, 135, 498-505.
- Zhou, Y.; Ren, Y.; Zhang, L.; You, L.; Yuan, Y.; Anslyn, E. V. Dynamic covalent binding and chirality sensing of mono secondary amines with a metal-templated assembly. Tetrahedron 2015, 71 (21), 3515-3521.