Isopentylamine, 99%
Isopentylamine, 99%
Isopentylamine, 99%
Thermo Scientific Chemicals
Isopentylamine, 99%
CAS: 107-85-7 | C5H13N | 87.166 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02260.1425 g
Catalog number ALFL02260.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10038-98-9
IUPAC Namegermanium tetrachloride
Molecular FormulaCl4GeH8
InChI KeyVDHBHZZBEGQJRK-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[GeH4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Total trace metal impurities=<0.01 %