Phenanthrene, 98%, Thermo Scientific Chemicals
Phenanthrene, 98%, Thermo Scientific Chemicals
Phenanthrene, 98%, Thermo Scientific Chemicals
Phenanthrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenanthrene, 98%, Thermo Scientific Chemicals
CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ALFL01921.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight178.23
Unit Size
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.
Solubility
Soluble in Ether, Benzene, Chloroform, Toluene. Insoluble in water.
Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. T. Azumi and S. P. McGlynn. Polarization of the Luminescence of Phenanthrene.J. Chem. Phys.1962, 37 2413.
  2. Yanzheng Gao; Lizhong Zhu. Plant uptake, accumulation and translocation of phenanthrene and pyrene in soils.Chemosphere.2004, 55 1169-1178.