Acridine, 97%, Thermo Scientific Chemicals
Acridine, 97%, Thermo Scientific Chemicals
Acridine, 97%, Thermo Scientific Chemicals
Acridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acridine, 97%, Thermo Scientific Chemicals
CAS: 260-94-6 | C13H9N | 179.222 g/mol
Have Questions?
Catalog NumberQuantity
ALFL01657.1425 g
Catalog number ALFL01657.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS530-62-1
IUPAC Name1-(1H-imidazole-1-carbonyl)-1H-imidazole
Molecular FormulaC7H6N4O
InChI KeyPFKFTWBEEFSNDU-UHFFFAOYSA-N
SMILESO=C(N1C=CN=C1)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (total imidazole content)
Solution Test5% solution in DMF is clear
Appearance (Color)White to cream