2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
CAS: 90-00-6 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFL01637.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS95201-93-7
IUPAC Name(2Z)-4-bromo-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Molecular FormulaC6H5BrO3S
InChI KeyYCTROWLDGFWISA-WAYWQWQTSA-N
SMILESCOC(O)=C1/SC=C(Br)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale orange to pale brown or pink/brown to pink/red
FormPowder
Melting Point (clear melt)76-82?C
Assay (GC)≥96.0%