9-Heptadecanone, 98%, Thermo Scientific Chemicals
9-Heptadecanone, 98%, Thermo Scientific Chemicals
9-Heptadecanone, 98%, Thermo Scientific Chemicals
9-Heptadecanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Heptadecanone, 98%, Thermo Scientific Chemicals
CAS: 540-08-9 | C17H34O | 254.458 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL01630.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material9-Heptadecanone
CAS540-08-9
Melting Point51°C to 53°C
Recommended StorageAmbient temperatures
RTECS NumberMI3925000
View more
9-Heptadecanone is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Heptadecanone is used as pharmaceutical intermediates.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Avoid excessive heat and light. Always store away from incompatible compounds such as oxidizing agents.
RUO – Research Use Only