2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Dinitrotoluene, 97%, Thermo Scientific Chemicals
CAS: 606-20-2 | C7H6N2O4 | 182.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL00856.06
Price (MYR)
285.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
285.00
Add to cart
Specifications
Chemical Name or Material2,6-Dinitrotoluene
CAS606-20-2
Health Hazard 1H301+H311+H331-H341-H350-H361f-H373
Health Hazard 2GHS H Statement
H301-H311-H331-H350-H341-H361-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c