4-Nitrodiphenylamine, 98+%, Thermo Scientific Chemicals
4-Nitrodiphenylamine, 98+%, Thermo Scientific Chemicals
4-Nitrodiphenylamine, 98+%, Thermo Scientific Chemicals
4-Nitrodiphenylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrodiphenylamine, 98+%, Thermo Scientific Chemicals
CAS: 836-30-6 | C12H10N2O2 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFL00728.03
Price (MYR)
473.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
473.00
Add to cart
Specifications
Chemical Name or Material4-Nitrodiphenylamine
CAS836-30-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point132°C to 135°C
View more
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
  1. J Hine.; M Hine. The Relative Acidity of Water, Methanol and Other Weak Acids in Isopropyl Alcohol Solution1 Journal of the American Chemical Society. 195274,(21) , 5266-5271.
  2. AA Banerjee.; D Mukesh. Heterogeneous catalytic transfer hydrogenation of 4-nitrodiphenylamine to p-phenylenediamines. Journal of the American Chemical Society. 19881 (18) , 1275-1276.
  3. Hammett indicator. Intermediate in the production of rubber antioxidants.