N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
CAS: 100-23-2 | C8H10N2O2 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFL00404.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Dimethyl-4-nitroaniline
CAS100-23-2
Health Hazard 1H301+H311+H331-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
View more
It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.
Solubility
Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Oleg Ya Borbulevych, et al. Experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.J. Mol. Struct.,2002,604,(1), 73-86.
  2. M. A. Ab Rani, et al. Understanding the polarity of ionic liquids.Phys. Chem. Chem. Phys.,2011,13,(37), 16831-16840.